Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289757.1 | NP_001276686.1 | 7 - 8 | 848 | 86 | |
NM_021470.6 | NP_067445.3 | 7 - 8 | 820 | 86 | ||
GenBank mRNA | AF190664.1 | - | 4 - 5 | 582 | 86 | |
AK005795.1 | - | 7 - 8 | 775 | 86 | ||
AK006379.1 | - | 7 - 8 | 764 | 86 | ||
AK016026.1 | - | 7 - 8 | 791 | 86 | ||
BC050964.1 | - | 7 - 8 | 763 | 86 | ||
BC061103.1 | - | 7 - 8 | 589 | 86 | ||
BC089493.1 | - | 7 - 8 | 763 | 6741898 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |