Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023670.3 | NP_076159.3 | 11 - 12 | 1804 | 66 | |
GenBank mRNA | AB046173.1 | - | 11 - 12 | 1321 | 66 | |
AK011689.1 | - | 11 - 12 | 66 | |||
AK045244.1 | - | 1 - 2 | 514 | 66 | ||
AK088465.1 | - | 11 - 12 | 1563 | 66 | ||
AK147140.1 | - | 11 - 12 | 1808 | 66 | ||
BC045138.1 | - | 11 - 12 | 1434 | 6306448 | 66 | |
BC049082.1 | - | 11 - 12 | 1470 | 5706238 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |