Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025645.2 | NP_079921.2 | 3 - 4 | 441 | 85 | |
GenBank mRNA | AK002371.1 | - | 3 - 4 | 405 | 85 | |
AK004277.1 | - | 3 - 4 | 371 | 85 | ||
AK136135.1 | - | 3 - 4 | 371 | 85 | ||
AK148443.1 | - | 3 - 4 | 380 | 85 | ||
AK152287.1 | - | 3 - 4 | 405 | 85 | ||
AK152352.1 | - | 3 - 4 | 399 | 85 | ||
AK169605.1 | - | 3 - 4 | 387 | 85 | ||
BC058365.1 | - | 3 - 4 | 373 | 5714866 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |