Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Get4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026269.2 | NP_080545.2 | 1 - 2 | 290 | 65 | |
GenBank mRNA | AK003551.1 | - | 1 - 2 | 239 | 65 | |
CJ053487.1 | - | 1 - 2 | 291 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |