Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026643.5 | NP_080919.3 | 2 - 3 | 84 | 79 | |
NM_029132.2 | NP_083408.1 | 2 - 3 | 152 | 79 | ||
GenBank mRNA | AF397908.1 | - | 2 - 3 | 80 | 79 | |
AK010530.1 | - | 2 - 3 | 103 | 79 | ||
BC138230.1 | - | 2 - 3 | 87 | 8861249 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |