Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100005842.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005842.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005842.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005838.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005838.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005838.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005835.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005835.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005835.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005829.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005829.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005829.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005840.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005840.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005840.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005837.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005837.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005837.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005839.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005839.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005839.mm.1 |
TC0100000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005841.mm.1 |
NM_026713.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005841.mm.1 |
NM_026713.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005841.mm.1 |
TC0100000681.mm.1 |
MTA TC |