Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083895.3 | NP_001077364.2 | 2 - 3 | 566 | 90 | |
NM_026838.4 | NP_081114.2 | 3 - 4 | 618 | 90 | ||
GenBank mRNA | AF393640.1 | - | 1 - 2 | 136 | 90 | |
AK136935.1 | - | 2 - 3 | 421 | 90 | ||
BC028307.1 | - | 3 - 4 | 347 | 3488526 | 90 | |
CN539245.1 | - | 3 - 4 | 381 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |