Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarres2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027852.2 | NP_082128.1 | 2 - 3 | 286 | 77 | |
GenBank mRNA | AK002298.1 | - | 2 - 3 | 287 | 77 | |
BC038914.1 | - | 2 - 3 | 306 | 5100583 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |