Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
NM_027852.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
AK002298.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
BC015275.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
BC038914.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
NM_027852.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
AK002298.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
BC015275.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P466147 |
BC038914.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
907730 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
317590 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
146862 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114157 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
NM_027852.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
AK002298.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
BC015275.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
BC038914.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
NM_027852.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
AK002298.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
BC015275.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P466147 |
BC038914.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428110 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428077 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428118 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428105 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428085 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428098 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428113 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428120 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428080 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428082 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428081 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428121 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428107 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428096 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428079 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428119 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428106 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428084 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428093 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428102 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428095 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428089 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428099 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428090 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24428086 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P350311 |
BP765942.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P350311 |
AK171463.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P350311 |
BC027309.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P350311 |
AK143222.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P350311 |
AK170792.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
NM_027852.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
AK002298.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
BC015275.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
BC038914.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
NM_027852.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
AK002298.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
BC015275.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P466147 |
BC038914.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104651 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05235139 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05553667 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05553669 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05235140 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05235136 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05235141 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05235144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24428090 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428096 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428107 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428093 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428125 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428113 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05849511 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05342429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428082 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428073 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428104 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05235137 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428126 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428118 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428076 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428117 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428071 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428115 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428112 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428075 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428098 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05130454 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428103 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428099 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05235140 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428108 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428124 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05769474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05782411 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428102 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428090 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428086 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428081 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428105 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428119 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428110 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428123 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05527672 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24428078 |
- |
- |