Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Telo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163661.1 | NP_001157133.1 | 17 - 18 | 2699 | 103 | |
NM_027880.2 | NP_082156.2 | 17 - 18 | 2610 | 103 | ||
GenBank mRNA | AK004582.1 | - | 17 - 18 | 2480 | 103 | |
AK028658.1 | - | 17 - 18 | 2464 | 103 | ||
AK050773.1 | - | 17 - 18 | 2429 | 103 | ||
AK143827.1 | - | 17 - 18 | 2700 | 103 | ||
AK146963.1 | - | 17 - 18 | 2486 | 103 | ||
AK154727.1 | - | 17 - 18 | 2512 | 103 | ||
AK158181.1 | - | 4 - 5 | 103 | |||
BC011077.1 | - | 17 - 18 | 2460 | 3984911 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |