Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028680.3 | NP_082956.2 | 9 - 10 | 1089 | 80 | |
GenBank mRNA | AF274590.1 | - | 9 - 10 | 1107 | 80 | |
AK040328.1 | - | 6 - 7 | 660 | 80 | ||
AK047217.1 | - | 9 - 10 | 1089 | 80 | ||
BC138143.1 | - | 9 - 10 | 1063 | 8861162 | 80 | |
BC138144.1 | - | 9 - 10 | 1063 | 8861163 | 80 | |
DQ266091.1 | - | 9 - 10 | 1130 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |