Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029787.2 | NP_084063.1 | 5 - 6 | 537 | 90 | |
GenBank mRNA | AF332059.1 | - | 5 - 6 | 461 | 90 | |
AF332060.1 | - | 5 - 6 | 461 | 90 | ||
AK002640.1 | - | 5 - 6 | 509 | 90 | ||
AK010858.1 | - | 5 - 6 | 514 | 90 | ||
AK128917.1 | - | 5 - 6 | 531 | 90 | ||
AK181262.1 | - | 1 - 2 | 35 | 90 | ||
AK183532.1 | - | 1 - 2 | 35 | 90 | ||
AK185013.1 | - | 1 - 2 | 36 | 89 | ||
AK190014.1 | - | 1 - 2 | 35 | 90 | ||
AK211940.1 | - | 1 - 2 | 35 | 90 | ||
AK211950.1 | - | 1 - 2 | 35 | 90 | ||
AK213922.1 | - | 1 - 2 | 35 | 90 | ||
BC004760.1 | - | 5 - 6 | 544 | 3154531 | 90 | |
BC032013.1 | - | 5 - 6 | 519 | 4238275 | 90 | |
BC043074.1 | - | 5 - 6 | 537 | 6404746 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |