Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033606.3 | NP_291084.1 | 10 - 11 | 2181 | 84 | |
GenBank mRNA | AF318278.1 | - | 9 - 10 | 1825 | 84 | |
BC048936.1 | - | 5 - 6 | 844 | 84 | ||
BC075680.1 | - | 10 - 11 | 2181 | 30612145 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |