Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpnmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053110.4 | NP_444340.3 | 3 - 4 | 574 | 95 | |
GenBank mRNA | AF322054.1 | - | 3 - 4 | 95 | ||
AJ251685.1 | - | 3 - 4 | 459 | 95 | ||
AK044764.1 | - | 3 - 4 | 95 | |||
AK076347.1 | - | 3 - 4 | 443 | 95 | ||
AK079220.1 | - | 416 | 95 | |||
AK149700.1 | - | 3 - 4 | 443 | 95 | ||
AK149740.1 | - | 3 - 4 | 481 | 95 | ||
AK150739.1 | - | 3 - 4 | 443 | 95 | ||
AK154617.1 | - | 3 - 4 | 478 | 95 | ||
AK159165.1 | - | 3 - 4 | 473 | 95 | ||
AK159270.1 | - | 3 - 4 | 481 | 95 | ||
AK159747.1 | - | 3 - 4 | 443 | 95 | ||
AK159908.1 | - | 3 - 4 | 449 | 95 | ||
AK170506.1 | - | 3 - 4 | 473 | 95 | ||
AK171347.1 | - | 3 - 4 | 449 | 95 | ||
AK171441.1 | - | 3 - 4 | 443 | 95 | ||
AK171465.1 | - | 3 - 4 | 443 | 95 | ||
AK171474.1 | - | 3 - 4 | 443 | 95 | ||
AK171517.1 | - | 3 - 4 | 561 | 95 | ||
AK171608.1 | - | 3 - 4 | 443 | 95 | ||
AK171618.1 | - | 3 - 4 | 443 | 95 | ||
AK209686.1 | - | 1 - 2 | 110 | 95 | ||
BC026375.1 | - | 3 - 4 | 4164706 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |