Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080448.4 | NP_536696.4 | 21 - 22 | 3652 | 100 | |
GenBank mRNA | AF338472.1 | - | 20 - 21 | 2810 | 100 | |
AF496547.1 | - | 21 - 22 | 2882 | 100 | ||
CF744803.1 | - | 2 - 3 | 444 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Mus musculus Probe spans exons |