Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133251.1 | NP_573514.1 | 4 - 5 | 530 | 77 | |
GenBank mRNA | AB036708.1 | - | 4 - 5 | 530 | 77 | |
AK146118.1 | - | 3 - 4 | 401 | 77 | ||
BC144928.1 | - | 4 - 5 | 478 | 9053476 | 77 | |
BC145949.1 | - | 3 - 4 | 462 | 40131252 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |