Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177409.3 | NP_803128.1 | 8 - 9 | 844 | 70 | |
GenBank mRNA | AK082751.1 | - | 8 - 9 | 845 | 70 | |
AY029530.1 | - | 8 - 9 | 781 | 70 | ||
BC018212.1 | - | 8 - 9 | 829 | 3980004 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |