Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100037119.mm.1 |
NM_001161329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037119.mm.1 |
NM_001161329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037119.mm.1 |
NM_133702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037119.mm.1 |
NM_133702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037119.mm.1 |
TC1100003945.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037120.mm.1 |
NM_001161329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037120.mm.1 |
NM_001161329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037120.mm.1 |
NM_133702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037120.mm.1 |
NM_133702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037120.mm.1 |
TC1100003945.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037118.mm.1 |
NM_001161329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037118.mm.1 |
NM_001161329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037118.mm.1 |
NM_133702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037118.mm.1 |
NM_133702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037118.mm.1 |
TC1100003945.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037121.mm.1 |
NM_001161329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037121.mm.1 |
NM_001161329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037121.mm.1 |
NM_133702.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037121.mm.1 |
NM_133702.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037121.mm.1 |
TC1100003945.mm.1 |
MTA TC |