Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400032193.mm.1 |
TC0400003885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032193.mm.1 |
NM_001163256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032193.mm.1 |
NM_001163256.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032198.mm.1 |
TC0400003885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032198.mm.1 |
NM_133754.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032198.mm.1 |
NM_133754.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032179.mm.1 |
TC0400003885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032179.mm.1 |
NM_001163256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032179.mm.1 |
NM_001163256.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032179.mm.1 |
NM_133754.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032179.mm.1 |
NM_133754.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032180.mm.1 |
TC0400003885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032180.mm.1 |
NM_001163256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032180.mm.1 |
NM_001163256.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032180.mm.1 |
NM_133754.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032180.mm.1 |
NM_133754.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032183.mm.1 |
TC0400003885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032183.mm.1 |
NM_001163256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032183.mm.1 |
NM_001163256.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032183.mm.1 |
NM_133754.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032183.mm.1 |
NM_133754.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032194.mm.1 |
TC0400003885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032194.mm.1 |
NM_001163256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032194.mm.1 |
NM_001163256.1 |
Refseq |