Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133763.1 | NP_598524.1 | 6 - 7 | 535 | 72 | |
GenBank mRNA | AB045974.1 | - | 6 - 7 | 528 | 72 | |
AK018299.1 | - | 6 - 7 | 562 | 72 | ||
BC003486.1 | - | 6 - 7 | 535 | 2811159 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |