Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113362.1 | NP_001106833.1 | 11 - 12 | 1768 | 67 | |
NM_001113364.1 | NP_001106835.1 | 8 - 9 | 882 | 67 | ||
NM_133910.3 | NP_598671.3 | 10 - 11 | 1587 | 67 | ||
GenBank mRNA | AB064521.1 | - | 8 - 9 | 745 | 67 | |
AB093293.1 | - | 9 - 10 | 1381 | 67 | ||
AK013092.1 | - | 8 - 9 | 808 | 67 | ||
AK029986.1 | - | 11 - 12 | 1768 | 67 | ||
BC011420.1 | - | 8 - 9 | 760 | 67 | ||
BC042515.1 | - | 10 - 11 | 1563 | 4020510 | 67 | |
BC096446.1 | - | 10 - 11 | 1579 | 6307706 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |