Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199274.1 | NP_001186203.1 | 2 - 3 | 598 | 82 | |
NM_134017.2 | NP_598778.1 | 2 - 3 | 402 | 82 | ||
GenBank mRNA | AK075600.1 | - | 2 - 3 | 303 | 82 | |
AK075947.1 | - | 2 - 3 | 337 | 82 | ||
AK153126.1 | - | 2 - 3 | 358 | 82 | ||
AK162087.1 | - | 2 - 3 | 337 | 82 | ||
BC003457.1 | - | 2 - 3 | 333 | 2648452 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |