Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134020.1 | NP_598781.1 | 1 - 2 | 164 | 65 | |
GenBank mRNA | AF368921.1 | - | 1 - 2 | 195 | 65 | |
AK078425.1 | - | 1 - 2 | 175 | 65 | ||
AK158876.1 | - | 1 - 2 | 197 | 65 | ||
BC023041.1 | - | 1 - 2 | 164 | 5360236 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |