Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134023.1 | NP_598784.1 | 2 - 3 | 373 | 75 | |
GenBank mRNA | AK140872.1 | - | 2 - 3 | 385 | 75 | |
AK153677.1 | - | 355 | 75 | |||
AK220243.1 | - | 384 | 75 | |||
BC018300.1 | - | 2 - 3 | 373 | 4193046 | 75 | |
BC037230.1 | - | 2 - 3 | 356 | 2650923 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |