Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170430.1 | NP_001163901.1 | 3 - 4 | 535 | 103 | |
NM_001170431.1 | NP_001163902.1 | 3 - 4 | 670 | 103 | ||
NM_134060.4 | NP_598821.1 | 3 - 4 | 513 | 103 | ||
GenBank mRNA | AK076592.1 | - | 3 - 4 | 508 | 103 | |
AK077208.1 | - | 3 - 4 | 489 | 103 | ||
AK141280.1 | - | 3 - 4 | 671 | 103 | ||
BC006881.1 | - | 3 - 4 | 527 | 3596363 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |