Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170430.1 | NP_001163901.1 | 7 - 8 | 993 | 56 | |
NM_001170431.1 | NP_001163902.1 | 7 - 8 | 1128 | 56 | ||
NM_134060.4 | NP_598821.1 | 7 - 8 | 971 | 56 | ||
GenBank mRNA | AK076592.1 | - | 7 - 8 | 966 | 56 | |
AK077208.1 | - | 7 - 8 | 947 | 56 | ||
AK141280.1 | - | 7 - 8 | 1129 | 56 | ||
BC006881.1 | - | 7 - 8 | 985 | 3596363 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |