Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164743.1 | NP_001158215.1 | 7 - 8 | 1138 | 84 | |
NM_134069.3 | NP_598830.2 | 8 - 9 | 1262 | 84 | ||
GenBank mRNA | AK128896.1 | - | 8 - 9 | 1246 | 84 | |
AK143896.1 | - | 8 - 9 | 1234 | 84 | ||
BC018252.1 | - | 8 - 9 | 1191 | 4240254 | 84 | |
BC024459.1 | - | 8 - 9 | 1218 | 4975341 | 84 | |
BC034179.1 | - | 8 - 9 | 1082 | 4972500 | 84 | |
BC034183.1 | - | 8 - 9 | 1067 | 4973620 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |