Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166456.1 | NP_001159928.1 | 7 - 8 | 868 | 76 | |
NM_001166458.1 | NP_001159930.1 | 7 - 8 | 1075 | 76 | ||
NM_134086.4 | NP_598847.2 | 7 - 8 | 1561 | 76 | ||
GenBank mRNA | AF184240.2 | - | 6 - 7 | 653 | 76 | |
AK029189.1 | - | 7 - 8 | 738 | 76 | ||
AK034130.1 | - | 7 - 8 | 1563 | 76 | ||
AK035098.1 | - | 6 - 7 | 580 | 76 | ||
AK047459.1 | - | 7 - 8 | 1092 | 76 | ||
AK049173.1 | - | 7 - 8 | 775 | 76 | ||
AK049294.1 | - | 7 - 8 | 841 | 76 | ||
AK050914.1 | - | 7 - 8 | 776 | 76 | ||
AK081724.1 | - | 7 - 8 | 1077 | 76 | ||
AK162612.1 | - | 7 - 8 | 926 | 76 | ||
AK163914.1 | - | 2 - 3 | 179 | 76 | ||
AK165188.1 | - | 7 - 8 | 1128 | 76 | ||
BC030378.1 | - | 7 - 8 | 777 | 5375319 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |