Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166456.1 | NP_001159928.1 | 14 - 15 | 1564 | 119 | |
NM_001166458.1 | NP_001159930.1 | 14 - 15 | 1771 | 119 | ||
NM_134086.4 | NP_598847.2 | 14 - 15 | 2257 | 119 | ||
GenBank mRNA | AF184240.2 | - | 13 - 14 | 1349 | 119 | |
AK034130.1 | - | 14 - 15 | 2259 | 119 | ||
AK035098.1 | - | 13 - 14 | 1276 | 119 | ||
AK047459.1 | - | 14 - 15 | 1788 | 119 | ||
AK049173.1 | - | 14 - 15 | 1473 | 119 | ||
AK049294.1 | - | 14 - 15 | 1537 | 119 | ||
AK081724.1 | - | 14 - 15 | 1773 | 119 | ||
AK082683.1 | - | 14 - 15 | 1475 | 119 | ||
AK162612.1 | - | 14 - 15 | 1622 | 119 | ||
AK163914.1 | - | 9 - 10 | 875 | 119 | ||
AK165188.1 | - | 14 - 15 | 1824 | 119 | ||
BC030378.1 | - | 14 - 15 | 1473 | 5375319 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |