Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083902.1 | NP_001077371.1 | 10 - 11 | 1782 | 136 | |
NM_134251.2 | NP_599012.2 | 11 - 12 | 1898 | 136 | ||
GenBank mRNA | AF319951.1 | - | 11 - 12 | 1892 | 136 | |
AK087781.1 | - | 7 - 8 | 1488 | 192 | ||
AK165080.1 | - | 6 - 7 | 1405 | 136 | ||
BC030926.1 | - | 10 - 11 | 1782 | 4910321 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |