Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134420.4 | NP_599252.2 | 960 | 79 | ||
GenBank mRNA | AB099881.1 | - | 121 | 79 | ||
AF248494.1 | - | 306 | 79 | |||
AK029238.1 | - | 1886 | 79 | |||
AK046993.1 | - | 1871 | 79 | |||
AK053088.1 | - | 262 | 79 | |||
AK080750.1 | - | 319 | 79 | |||
AK163670.1 | - | 961 | 79 | |||
AK163671.1 | - | 961 | 79 | |||
AY032863.1 | - | 265 | 79 | |||
AY049076.1 | - | 220 | 79 | |||
BC028856.1 | - | 284 | 4165725 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |