Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138582.1 | NP_613048.1 | 1 - 2 | 188 | 87 | |
GenBank mRNA | AF134319.1 | - | 1 - 2 | 188 | 87 | |
BC137792.1 | - | 1 - 2 | 129 | 8860811 | 87 | |
BC144911.1 | - | 1 - 2 | 129 | 9053459 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |