Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138628.3 | NP_619534.2 | 8 - 9 | 1389 | 101 | |
GenBank mRNA | AF422244.1 | - | 8 - 9 | 1326 | 101 | |
AF422245.1 | - | 8 - 9 | 1326 | 101 | ||
AK084639.1 | - | 8 - 9 | 1390 | 101 | ||
AK085904.1 | - | 8 - 9 | 1285 | 101 | ||
AK137442.1 | - | 8 - 9 | 1304 | 101 | ||
BC141023.1 | - | 8 - 9 | 1302 | 9055635 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |