Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138657.3 | NP_619598.1 | 3 - 4 | 1037 | 58 | |
GenBank mRNA | AF424814.1 | - | 3 - 4 | 1038 | 58 | |
AK147580.1 | - | 3 - 4 | 1135 | 58 | ||
AK147650.1 | - | 3 - 4 | 1135 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |