Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138683.2 | NP_619624.2 | 4 - 5 | 920 | 95 | |
GenBank mRNA | AB016768.1 | - | 4 - 5 | 920 | 95 | |
AK132277.1 | - | 4 - 5 | 867 | 95 | ||
BC138831.1 | - | 3 - 4 | 672 | 8861853 | 95 | |
BC138833.1 | - | 3 - 4 | 672 | 8861855 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |