Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139001.2 | NP_620570.2 | 3 - 4 | 3893 | 120 | |
GenBank mRNA | AF352400.1 | - | 3 - 4 | 3837 | 120 | |
AK052394.1 | - | 3 - 4 | 3965 | 120 | ||
AK220559.1 | - | 1 - 2 | 814 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |