Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145221.2 | NP_660256.1 | 20 - 21 | 1986 | 104 | |
GenBank mRNA | AK030445.1 | - | 20 - 21 | 1865 | 104 | |
AK033566.1 | - | 20 - 21 | 1957 | 104 | ||
AK045438.1 | - | 20 - 21 | 1922 | 104 | ||
AK146356.1 | - | 20 - 21 | 1957 | 104 | ||
AK173169.1 | - | 5 - 6 | 437 | 104 | ||
AY113705.1 | - | 20 - 21 | 1951 | 104 | ||
BC019708.1 | - | 12 - 13 | 1264 | 104 | ||
BC063751.1 | - | 20 - 21 | 1998 | 6511401 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |