Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291144.1 | NP_001278073.1 | 1 - 2 | 122 | 91 | |
NM_145742.2 | NP_665685.1 | 1 - 2 | 122 | 91 | ||
GenBank mRNA | AK004665.1 | - | 1 - 2 | 48 | 91 | |
AK155482.1 | - | 1 - 2 | 66 | 91 | ||
AK156955.1 | - | 1 - 2 | 68 | 91 | ||
AK170952.1 | - | 1 - 2 | 66 | 91 | ||
BC029709.1 | - | 1 - 2 | 109 | 5359253 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |