Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB089793.1 | - | 3 - 4 | 309 | 103 | |
AF482993.1 | - | 2 - 3 | 273 | 103 | ||
AK037377.1 | - | 3 - 4 | 296 | 103 | ||
AK049410.1 | - | 3 - 4 | 305 | 103 | ||
AK143281.1 | - | 3 - 4 | 278 | 103 | ||
AK160826.1 | - | 3 - 4 | 284 | 103 | ||
AK173069.1 | - | 3 - 4 | 645 | 103 | ||
BC030879.1 | - | 3 - 4 | 308 | 4241988 | 103 | |
BC058947.1 | - | 3 - 4 | 279 | 5702781 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |