Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152823.4 | NP_690036.4 | 7 - 8 | 1355 | 100 | |
GenBank mRNA | AK039924.1 | - | 3 - 4 | 285 | 100 | |
AK089034.1 | - | 3 - 4 | 100 | |||
AK144925.1 | - | 7 - 8 | 1356 | 100 | ||
AY510108.1 | - | 6 - 7 | 1222 | 100 | ||
BC022701.1 | - | 3 - 4 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |