Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102455.1 | NP_001095925.1 | 1 - 2 | 191 | 106 | |
NM_001102456.1 | NP_001095926.1 | 1 - 2 | 191 | 106 | ||
NM_009691.2 | NP_033821.1 | 1 - 2 | 191 | 106 | ||
GenBank mRNA | AK013376.1 | - | 1 - 2 | 188 | 106 | |
AK149977.1 | - | 1 - 2 | 192 | 106 | ||
AK150020.1 | - | 1 - 2 | 192 | 106 | ||
AK160471.1 | - | 1 - 2 | 191 | 106 | ||
AK164012.1 | - | 1 - 2 | 209 | 106 | ||
BC051999.1 | - | 1 - 2 | 169 | 5716148 | 106 | |
BC052396.1 | - | 1 - 2 | 163 | 5719918 | 106 | |
U15571.1 | - | 1 - 2 | 167 | 106 | ||
Z22592.1 | - | 1 - 2 | 191 | 106 | ||
Z27070.1 | - | 1 - 2 | 189 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |