Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020033.1 | NP_064417.1 | 2 - 3 | 189 | 69 | |
GenBank mRNA | AB076256.1 | - | 2 - 3 | 201 | 69 | |
AJ011118.1 | - | 2 - 3 | 189 | 69 | ||
AJ245514.1 | - | 2 - 3 | 189 | 69 | ||
AK132585.1 | - | 2 - 3 | 199 | 69 | ||
BC138760.1 | - | 2 - 3 | 227 | 8861782 | 69 | |
BC138761.1 | - | 2 - 3 | 227 | 8861783 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |