Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021535.4 | NP_067510.3 | 10 - 11 | 1372 | 71 | |
GenBank mRNA | AB044414.1 | - | 10 - 11 | 1301 | 71 | |
AK003493.1 | - | 10 - 11 | 1359 | 71 | ||
AK019174.1 | - | 10 - 11 | 1331 | 71 | ||
AK145982.1 | - | 10 - 11 | 1371 | 71 | ||
AK159635.1 | - | 10 - 11 | 1359 | 71 | ||
AK168813.1 | - | 10 - 11 | 1366 | 71 | ||
AK212788.1 | - | 1 - 2 | 177 | 71 | ||
BC057446.1 | - | 10 - 11 | 1333 | 3155760 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |