Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024462.2 | NP_077782.1 | 1 - 2 | 178 | 57 | |
GenBank mRNA | AK010418.1 | - | 1 - 2 | 108 | 57 | |
AK079783.1 | - | 1 - 2 | 37 | 57 | ||
BC002274.1 | - | 1 - 2 | 91 | 3496493 | 57 | |
BF012097.1 | - | 1 - 2 | 182 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |