Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025616.3 | NP_079892.1 | 3 - 4 | 297 | 88 | |
GenBank mRNA | AK008340.1 | - | 3 - 4 | 298 | 88 | |
AK008382.1 | - | 3 - 4 | 295 | 88 | ||
AK010832.1 | - | 3 - 4 | 263 | 88 | ||
AK012977.1 | - | 3 - 4 | 294 | 88 | ||
AK088074.1 | - | 3 - 4 | 290 | 88 | ||
AK167473.1 | - | 3 - 4 | 300 | 88 | ||
AK178698.1 | - | 2 - 3 | 71 | 88 | ||
AY551342.1 | - | 3 - 4 | 283 | 88 | ||
BC010303.1 | - | 3 - 4 | 272 | 3597676 | 88 | |
BC037056.1 | - | 3 - 4 | 299 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |