Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291054.1 | NP_001277983.1 | 11 - 12 | 1586 | 72 | |
NM_025676.4 | NP_079952.2 | 10 - 11 | 1502 | 72 | ||
GenBank mRNA | AK010365.1 | - | 2 - 3 | 224 | 72 | |
AK017607.1 | - | 11 - 12 | 1484 | 72 | ||
AK133858.1 | - | 11 - 12 | 1543 | 72 | ||
BC046780.1 | - | 11 - 12 | 1533 | 5716136 | 72 | |
BC052070.1 | - | 10 - 11 | 1308 | 5716125 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |