Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508729_s1
          See other RNF186 GE Assays ›
          Gene Symbol:
          Rnf186
          Gene Name:
          ring finger protein 186
          Gene Aliases:
          9130020G10Rik
          Chromosome Location:
          Chr.4: 138967112 - 138968366 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508729_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf186
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025786.3 NP_080062.4 1 242 79
          GenBank mRNA AI119068.1 - 1 242 79
          AK020617.1 - 1 151 79
          AK078919.1 - 1 237 79
          BC011492.1 - 1 207 4207836 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001737.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK078919.1,BC011492.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf186

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159934 BC011492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159934 CF379387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159934 NM_025786.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159934 AK078919.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315730 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137705 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159934 BC011492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159934 CF379387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159934 NM_025786.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159934 AK078919.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23348896 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137705 - -
          Whole Mouse Genome G4122A A_51_P272023 BC011492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272023 CF379387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272023 NM_025786.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272023 AK020617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272023 AK078919.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110476 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03733134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04046298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03733140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03733137 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04046297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03733127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04046300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03757853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03757862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03855593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03790341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03520168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04085135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03424783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03757858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03733148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509597 - -
          Affymetrix HT MG-430 PM Array Plate 1424794_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509596 - -
          Mouse Genome 430 2.0 Array 1424794_at - -
          Mouse Genome 430 2.0 Array 1424794_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66825_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66825_at - -
          Affymetrix Mouse Clariom S Array PSR0400014182.mm.1 NM_025786.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014182.mm.1 NM_025786.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014182.mm.1 TC0400001737.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014183.mm.1 NM_025786.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014183.mm.1 NM_025786.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014183.mm.1 TC0400001737.mm.1 MTA TC
          Mouse Expression Set 430 1424794_PM_at - -
          Mouse Expression Set 430 1424794_at - -
          Mouse Genome 430A 2.0 Array 1424794_PM_at - -
          Mouse Genome 430A 2.0 Array 1424794_at - -
          Affymetrix HT Mouse Genome 430A Array 1424794_at - -
          Affymetrix HT Mouse Genome 430A Array 1424794_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 334943 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 520286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2663996 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2663996 - -
          mouse-6 v1.1 (Illumina) scl24781.1.87_255-S - -
          Illumina Sentrix mouse-6.v1 bead array 520286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24781.1.87_255-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2663996 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663996 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24781.1.87_255-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0