Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025833.3 | NP_080109.1 | 1 - 2 | 323 | 72 | |
GenBank mRNA | AK004918.2 | - | 1 - 2 | 324 | 72 | |
AK008203.1 | - | 1 - 2 | 228 | 72 | ||
BC015459.1 | - | 1 - 2 | 101 | 3592632 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |