Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077348.1 | NP_001070816.1 | 5 - 6 | 739 | 76 | |
NM_025874.3 | NP_080150.2 | 6 - 7 | 685 | 76 | ||
GenBank mRNA | AK010202.1 | - | 6 - 7 | 685 | 76 | |
AK075872.1 | - | 6 - 7 | 682 | 76 | ||
AY919875.1 | - | 5 - 6 | 698 | 76 | ||
BC024138.1 | - | 6 - 7 | 665 | 5136320 | 76 | |
DQ473305.1 | - | 6 - 7 | 665 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |