Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286986.1 | NP_001273915.1 | 5 - 6 | 596 | 93 | |
NM_001286987.1 | NP_001273916.1 | 4 - 5 | 474 | 93 | ||
NM_026013.3 | NP_080289.1 | 5 - 6 | 599 | 93 | ||
GenBank mRNA | AK009940.1 | - | 5 - 6 | 536 | 93 | |
AK012044.1 | - | 5 - 6 | 516 | 93 | ||
AK015888.1 | - | 5 - 6 | 562 | 93 | ||
AK052824.1 | - | 4 - 5 | 408 | 93 | ||
AK172455.1 | - | 5 - 6 | 566 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |